Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001124914.1 | NP_001118386.1 | 5 - 6 | 564 | 68 |
NM_001336027.1 | NP_001325078.1 | 7 - 8 | 1855 | 68 | |
NM_001336028.1 | NP_001325077.1 | 8 - 9 | 1711 | 68 | |
NM_128148.4 | NP_180159.2 | 6 - 7 | 697 | 68 | |
GenBank mRNA | AK175188.1 | - | 6 - 7 | 661 | 68 |