Assay Design
Gene Transcripts
- Gene Symbol
- GLDP2
- Entrez Gene ID
-
817149
- Gene Name
- glycine decarboxylase P-protein 2
- Gene Aliases
-
AT2G26080, AtGLDP2, T19L18.11, T19L18_11, glycine decarboxylase P-protein 2
- Chromosome Location
-
Chr.2: 11109003 - 11114291 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_128167.4 |
NP_180178.1 |
1 - 2 |
1742 |
83 |
View Full Product Details