Assay Design
Gene Transcripts
- Gene Symbol
- ftsh4
- Entrez Gene ID
-
817154
- Gene Name
- FTSH protease 4
- Gene Aliases
-
AT2G26140, FTSH protease 4, T19L18.5, T19L18_5
- Chromosome Location
-
Chr.2: 11131578 - 11135465 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_128172.4 |
NP_565616.1 |
6 - 7 |
1569 |
78 |
View Full Product Details