Assay Design



Gene Transcripts

Gene Symbol
HO2
Entrez Gene ID
817196
Gene Name
heme oxygenase 2
Gene Aliases
AT2G26550, T9J22.22, T9J22_22, heme oxygenase 2
Chromosome Location
Chr.2: 11291148 - 11293575 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202681.2 NP_001189610.1 3 - 4 953 74
NM_001202682.1 NP_001189611.1 4 - 5 868 74
NM_128212.3 NP_180223.2 5 - 6 988 74

View Full Product Details