Assay Design
Gene Transcripts
- Gene Symbol
- SPX2
- Entrez Gene ID
-
817207
- Gene Name
- SPX domain-containing protein 2 (SPX2)
- Gene Aliases
-
ARABIDOPSIS THALIANA SPX DOMAIN GENE 2, AT2G26660, ATSPX2, F18A8.3, F18A8_3, SPX domain gene 2
- Chromosome Location
-
Chr.2: 11338752 - 11340950 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_128223.3 |
NP_180234.1 |
2 - 3 |
600 |
153 |
View Full Product Details