Assay Design



Gene Transcripts

Gene Symbol
RR13
Entrez Gene ID
817249
Gene Name
response regulator 13
Gene Aliases
ARR13, AT2G27070, T20P8.12, T20P8_12, response regulator 13
Chromosome Location
Chr.2: 11556392 - 11560215 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336100.1 NP_001318296.1 5 - 6 1080 70

View Full Product Details