Assay Design



Gene Transcripts

Gene Symbol
TIL2
Entrez Gene ID
817254
Gene Name
DNA polymerase epsilon catalytic subunit
Gene Aliases
AT2G27120, POL2B, T20P8.17, T20P8_17, TILTED 2
Chromosome Location
Chr.2: 11581214 - 11594397 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336102.1 NP_001325161.1 7 - 8 837 122
NM_001336103.1 NP_001325160.1 7 - 8 837 122
NM_128270.3 NP_180280.2 7 - 8 837 122

View Full Product Details