Assay Design



Gene Transcripts

Gene Symbol
AAO3
Entrez Gene ID
817257
Gene Name
abscisic aldehyde oxidase 3
Gene Aliases
ABSCISIC ALDEHYDE OXIDASE 3, AOdelta, AT2G27150, Aldehyde oxidase delta, Arabidopsis thaliana aldehyde oxidase 3, At-AO3, AtAAO3, F20F1.2, F20F1_2, abscisic aldehyde oxidase 3
Chromosome Location
Chr.2: 11601727 - 11608450 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084497.1 NP_001077966.1 1 - 2 324 131
NM_001336105.1 NP_001324270.1 1 - 2 346 131
NM_128273.3 NP_180283.1 1 - 2 351 131

View Full Product Details