Assay Design



Gene Transcripts

Gene Symbol
UXS6
Entrez Gene ID
817426
Gene Name
UDP-XYL synthase 6
Gene Aliases
AT2G28760, F8N16.5, F8N16_5, UDP-XYL synthase 6
Chromosome Location
Chr.2: 12336045 - 12339217 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084503.2 NP_001077972.1 6 - 7 655 109
NM_001336187.1 NP_001325413.1 6 - 7 752 109
NM_128436.5 NP_180443.1 6 - 7 782 109
NM_201826.3 NP_973555.1 6 - 7 855 109
GenBank mRNA AY099703.1 - 6 - 7 670 109
AY128899.1 - 5 - 6 586 109

View Full Product Details