Assay Design
Gene Transcripts
- Gene Symbol
- ASA2
- Entrez Gene ID
-
817519
- Gene Name
- anthranilate synthase 2
- Gene Aliases
-
AT2G29690, ATHANSYNAB, T27A16.21, T27A16_21, anthranilate synthase 2
- Chromosome Location
-
Chr.2: 12693734 - 12697091 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_128523.5 |
NP_180530.1 |
10 - 11 |
1669 |
91 |
View Full Product Details