Assay Design



Gene Transcripts

Gene Symbol
ABCC8
Entrez Gene ID
821679
Gene Name
multidrug resistance-associated protein 6
Gene Aliases
ARABIDOPSIS THALIANA MULTIDRUG RESISTANCE-ASSOCIATED PROTEIN 6, AT3G21250, ATMRP6, ATP-binding cassette C8, MRP6, multidrug resistance-associated protein 6
Chromosome Location
Chr.3: 7457433 - 7463644 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203015.2 NP_001189944.1 3 - 4 2667 92
NM_001338517.1 NP_001326627.1 4 - 5 2578 92
NM_001338518.1 NP_001326629.1 3 - 4 2557 92
NM_001338519.1 NP_001326630.1 3 - 4 2438 92
NM_001338520.1 NP_001326628.1 4 - 5 2349 92
NM_113020.3 NP_188762.3 3 - 4 2548 92

View Full Product Details