Assay Design



Gene Transcripts

Gene Symbol
ABCC8
Entrez Gene ID
821679
Gene Name
multidrug resistance-associated protein 6
Gene Aliases
ARABIDOPSIS THALIANA MULTIDRUG RESISTANCE-ASSOCIATED PROTEIN 6, AT3G21250, ATMRP6, ATP-binding cassette C8, MRP6, multidrug resistance-associated protein 6
Chromosome Location
Chr.3: 7457433 - 7463644 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203015.2 NP_001189944.1 7 - 8 3831 92
NM_001338517.1 NP_001326627.1 8 - 9 3742 92
NM_001338518.1 NP_001326629.1 7 - 8 3721 92
NM_001338519.1 NP_001326630.1 7 - 8 3569 92
NM_001338520.1 NP_001326628.1 8 - 9 3513 92
NM_113020.3 NP_188762.3 7 - 8 3679 92

View Full Product Details