Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001203015.2 | NP_001189944.1 | 10 - 11 | 4411 | 78 |
NM_001338517.1 | NP_001326627.1 | 11 - 12 | 4322 | 78 | |
NM_001338518.1 | NP_001326629.1 | 10 - 11 | 4301 | 78 | |
NM_001338519.1 | NP_001326630.1 | 10 - 11 | 4149 | 78 | |
NM_001338520.1 | NP_001326628.1 | 11 - 12 | 4093 | 78 | |
NM_113020.3 | NP_188762.3 | 10 - 11 | 4259 | 78 |