Assay Design



Gene Transcripts

Gene Symbol
UBP7
Entrez Gene ID
821682
Gene Name
ubiquitin carboxyl-terminal hydrolase
Gene Aliases
AT3G21280, ubiquitin-specific protease 7
Chromosome Location
Chr.3: 7477930 - 7482125 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338521.1 NP_001325430.1 13 - 14 1195 73
NM_113023.4 NP_566680.2 14 - 15 1448 73

View Full Product Details