Assay Design



Gene Transcripts

Gene Symbol
PMH1
Entrez Gene ID
821800
Gene Name
putative mitochondrial RNA helicase 1
Gene Aliases
AT3G22310, ATRH9, RNA HELICASE 9, putative mitochondrial RNA helicase 1
Chromosome Location
Chr.3: 7887276 - 7890153 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338579.1 NP_001327663.1 4 - 5 1165 141
NM_113129.4 NP_188870.1 4 - 5 1175 141
GenBank mRNA AY091091.1 - 4 - 5 1165 141

View Full Product Details