Assay Design



Gene Transcripts

Gene Symbol
NRPB5
Entrez Gene ID
821801
Gene Name
Eukaryotic rpb5 RNA polymerase subunit family protein
Gene Aliases
A, AT3G22320, ATRPABC24.3, NRPD5, RNA POLYMERASE II FIFTH LARGEST SUBUNIT, RPB5A
Chromosome Location
Chr.3: 7890803 - 7892349 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113130.4 NP_188871.1 3 - 4 526 108
GenBank mRNA AF019248.1 - 3 - 4 495 108
AF370551.1 - 3 - 4 500 108
AF412101.1 - 3 - 4 501 108
AY086150.1 - 3 - 4 500 108
BT002130.1 - 2 - 3 381 108

View Full Product Details