Assay Design
Gene Transcripts
- Gene Symbol
- LOX5
- Entrez Gene ID
-
821808
- Gene Name
- PLAT/LH2 domain-containing lipoxygenase family protein
- Gene Aliases
-
AT3G22400, ATLOX5, Arabidopsis thaliana lipoxygenase 5
- Chromosome Location
-
Chr.3: 7926728 - 7931351 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_113137.4 |
NP_188879.2 |
4 - 5 |
1414 |
67 |
View Full Product Details