Assay Design



Gene Transcripts

Gene Symbol
PAP85
Entrez Gene ID
821835
Gene Name
cupin family protein
Gene Aliases
AT3G22640
Chromosome Location
Chr.3: 8011724 - 8013941 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113163.4 NP_566714.1 3 - 4 1012 90
GenBank mRNA AY058085.1 - 3 - 4 973 90
AY090307.1 - 3 - 4 954 90

View Full Product Details