Assay Design



Gene Transcripts

Gene Symbol
HMT3
Entrez Gene ID
821845
Gene Name
homocysteine S-methyltransferase 3
Gene Aliases
AT3G22740, homocysteine S-methyltransferase 3
Chromosome Location
Chr.3: 8032917 - 8036005 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113173.3 NP_566715.1 6 - 7 1171 66
GenBank mRNA AF297394.1 - 6 - 7 876 66
AK118021.1 - 6 - 7 971 66
BT005318.1 - 6 - 7 876 66

View Full Product Details