Assay Design
Gene Transcripts
- Gene Symbol
- PHO1
- Entrez Gene ID
-
821924
- Gene Name
- phosphate 1
- Gene Aliases
-
ARABIDOPSIS PHOSPHATE 1, AT3G23430, ATPHO1, phosphate 1
- Chromosome Location
-
Chr.3: 8387398 - 8393467 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_113246.5 |
NP_188985.2 |
1 - 2 |
474 |
87 |
GenBank mRNA |
AF474076.1 |
- |
1 - 2 |
303 |
87 |
View Full Product Details