Assay Design



Gene Transcripts

Gene Symbol
PERK1
Entrez Gene ID
822051
Gene Name
proline extensin-like receptor kinase 1
Gene Aliases
AT3G24550, ATPERK1, MOB24.11, proline extensin-like receptor kinase 1, proline-rich extensin-like receptor kinase 1
Chromosome Location
Chr.3: 8960095 - 8963568 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113366.3 NP_189098.1 7 - 8 1957 96
GenBank mRNA AY056788.1 - 7 - 8 1677 96
AY059901.1 - 7 - 8 1672 96
AY093065.1 - 7 - 8 1670 96
AY128792.1 - 7 - 8 1641 96
BT008400.1 - 7 - 8 1641 96

View Full Product Details