Assay Design



Gene Transcripts

Gene Symbol
AFB2
Entrez Gene ID
822296
Gene Name
auxin signaling F-box 2
Gene Aliases
AT3G26810, auxin signaling F-box 2
Chromosome Location
Chr.3: 9867652 - 9870782 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113593.3 NP_566800.1 2 - 3 1633 99
GenBank mRNA AY062568.1 - 2 - 3 1444 99
BT010396.1 - 2 - 3 943 99

View Full Product Details