Assay Design
Gene Transcripts
- Gene Symbol
- CDG1
- Entrez Gene ID
-
822312
- Gene Name
- Protein kinase superfamily protein
- Gene Aliases
-
AT3G26940, CONSTITUTIVE DIFFERENTIAL GROWTH 1
- Chromosome Location
-
Chr.3: 9936582 - 9939305 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_113608.3 |
NP_189330.1 |
3 - 4 |
846 |
136 |
GenBank mRNA |
DQ446706.1 |
- |
3 - 4 |
477 |
136 |
View Full Product Details