Assay Design



Gene Transcripts

Gene Symbol
HA11
Entrez Gene ID
836388
Gene Name
H[+]-ATPase 11
Gene Aliases
AHA11, AT5G62670, H(+)-ATPase 11, MRG21.9, MRG21_9
Chromosome Location
Chr.5: 25159208 - 25165209 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125662.4 NP_201073.1 18 - 19 2395 66
GenBank mRNA AY125493.1 - 18 - 19 2371 66
BT010748.1 - 17 - 18 2225 66

View Full Product Details