Assay Design



Gene Transcripts

Gene Symbol
OPT4
Entrez Gene ID
836562
Gene Name
oligopeptide transporter 4
Gene Aliases
ARABIDOPSIS THALIANA OLIGOPEPTIDE TRANSPORTER 4, AT5G64410, ATOPT4, MSJ1.25, MSJ1_25, oligopeptide transporter 4
Chromosome Location
Chr.5: 25750845 - 25755501 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125837.4 NP_201246.1 3 - 4 1665 68
GenBank mRNA AK227021.1 - 3 - 4 1216 68
AY099843.1 - 3 - 4 1212 68
BT000339.1 - 3 - 4 1138 68

View Full Product Details