Assay Design
Gene Transcripts
- Gene Symbol
- FAAH
- Entrez Gene ID
-
836565
- Gene Name
- fatty acid amide hydrolase
- Gene Aliases
-
AT5G64440, AtFAAH, fatty acid amide hydrolase
- Chromosome Location
-
Chr.5: 25765712 - 25770618 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_125840.4 |
NP_201249.2 |
8 - 9 |
999 |
60 |
GenBank mRNA |
AY308736.1 |
- |
8 - 9 |
915 |
60 |
View Full Product Details