Assay Design
Gene Transcripts
- Gene Symbol
- CESA6
- Entrez Gene ID
-
836595
- Gene Name
- cellulose synthase 6
- Gene Aliases
-
AT5G64740, E112, ISOXABEN RESISTANT 2, IXR2, MVP7.7, MVP7_7, PRC1, PROCUSTE 1, cellulose synthase 6
- Chromosome Location
-
Chr.5: 25881033 - 25886728 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_125870.3 |
NP_201279.1 |
10 - 11 |
2643 |
78 |
GenBank mRNA |
KJ676983.1 |
- |
10 - 11 |
2121 |
78 |
View Full Product Details