Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037067.1 | NP_001032144.1 | 5 - 6 | 446 | 120 |
NM_001345663.1 | NP_001330809.1 | 5 - 6 | 667 | 120 | |
NM_125905.4 | NP_201311.1 | 5 - 6 | 667 | 120 | |
GenBank mRNA | AY231445.1 | - | 5 - 6 | 547 | 120 |
AY231446.1 | - | 5 - 6 | 416 | 120 | |
BT025758.1 | - | 5 - 6 | 416 | 120 |