Assay Design



Gene Transcripts

Gene Symbol
UBP17
Entrez Gene ID
836670
Gene Name
ubiquitin-specific protease 17
Gene Aliases
AT5G65450, MNA5.19, MNA5_19, ubiquitin-specific protease 17
Chromosome Location
Chr.5: 26157535 - 26161331 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345687.1 NP_001332735.1 6 - 7 1591 88
NM_125943.2 NP_201348.1 6 - 7 1263 88

View Full Product Details