Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161327.1 | NP_001154799.1 | 5 - 6 | 1110 | 156 |
NM_001203697.1 | NP_001190626.1 | 5 - 6 | 1305 | 156 | |
NM_001203698.1 | NP_001190627.1 | 5 - 6 | 1329 | 156 | |
NM_125974.5 | NP_201378.5 | 5 - 6 | 1029 | 156 |