Assay Design



Gene Transcripts

Gene Symbol
ATSAC1B
Entrez Gene ID
836732
Gene Name
Phosphoinositide phosphatase family protein
Gene Aliases
AT5G66020, ATSAC6, IBS2, IMPAIRED IN BABA-INDUCED STERILITY 2, K2A18.9, K2A18_9, SAC DOMAIN-CONTAINING PROTEIN 6, SACIB, SUPPRESSOR OF ACTIN 1B
Chromosome Location
Chr.5: 26401193 - 26404725 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125999.3 NP_201403.2 13 - 14 1143 78
GenBank mRNA AF266458.1 - 13 - 14 1109 78
AY227249.1 - 13 - 14 1109 78

View Full Product Details