Assay Design



Gene Transcripts

Gene Symbol
RFL1
Entrez Gene ID
837774
Gene Name
RPS5-like 1
Gene Aliases
AT1G12210, RPS5-like 1, T28K15.6, T28K15_6
Chromosome Location
Chr.1: 4140216 - 4143916 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332021.1 NP_001321385.1 2 - 2 2725 129
NM_001332022.1 NP_001321386.1 2 - 2 2687 129
NM_101093.2 NP_172685.1 1 - 1 2628 129

View Full Product Details