Assay Design
Gene Transcripts
- Gene Symbol
- AFP2
- Entrez Gene ID
-
837934
- Gene Name
- ABI five binding protein 2
- Gene Aliases
-
ABI five binding protein 2, AT1G13740, F21F23.17, F21F23_17
- Chromosome Location
-
Chr.1: 4713433 - 4715407 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_101242.3 |
NP_563933.1 |
1 - 2 |
1166 |
85 |
GenBank mRNA |
BT024887.1 |
- |
1 - 2 |
630 |
85 |
View Full Product Details