Assay Design
Gene Transcripts
- Gene Symbol
- VIK
- Entrez Gene ID
-
837960
- Gene Name
- VH1-interacting kinase
- Gene Aliases
-
AT1G14000, F7A19.9, F7A19_9, VH1-interacting kinase
- Chromosome Location
-
Chr.1: 4797358 - 4800283 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_101266.4 |
NP_172853.1 |
6 - 7 |
1048 |
97 |
GenBank mRNA |
BT026491.1 |
- |
6 - 7 |
800 |
97 |
View Full Product Details