Assay Design
Gene Transcripts
- Gene Symbol
- TWN2
- Entrez Gene ID
-
838023
- Gene Name
- valyl-tRNA synthetase / valine-tRNA ligase (VALRS)
- Gene Aliases
-
AT1G14610, T5E21.11, T5E21_11, TWIN 2, VALRS, VALYL TRNA SYNTHETASE
- Chromosome Location
-
Chr.1: 5008153 - 5014533 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_101328.4 |
NP_172913.1 |
10 - 11 |
958 |
151 |
View Full Product Details