Assay Design



Gene Transcripts

Gene Symbol
PBC1
Entrez Gene ID
838776
Gene Name
proteasome beta subunit C1
Gene Aliases
20S PROTEASOME BETA SUBUNIT PBC1, AT1G21720, F8K7.15, F8K7_15, proteasome beta subunit C1
Chromosome Location
Chr.1: 7626182 - 7628290 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102021.3 NP_564149.1 2 - 3 401 75
GenBank mRNA AK226397.1 - 2 - 3 277 75
AY065070.1 - 2 - 3 248 75
AY114628.1 - 1 - 2 189 75

View Full Product Details