Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001332527.1 | NP_001320240.1 | 9 - 10 | 1252 | 60 |
NM_102025.3 | NP_564150.1 | 9 - 10 | 1064 | 60 | |
GenBank mRNA | AK228033.1 | - | 9 - 10 | 984 | 60 |
BT006427.1 | - | 8 - 9 | 934 | 60 |