Assay Design



Gene Transcripts

Gene Symbol
IQD31
Entrez Gene ID
843808
Gene Name
IQ-domain 31
Gene Aliases
AT1G74690, F25A4.33, F25A4_33, IQ-domain 31
Chromosome Location
Chr.1: 28061159 - 28064771 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106127.5 NP_177607.2 4 - 5 900 89
GenBank mRNA AY099679.1 - 4 - 5 753 89
AY128860.1 - 3 - 4 582 89

View Full Product Details