Assay Design



Gene Transcripts

Gene Symbol
EDS16
Entrez Gene ID
843810
Gene Name
ADC synthase superfamily protein
Gene Aliases
ARABIDOPSIS ISOCHORISMATE SYNTHASE 1, AT1G74710, ATICS1, ENHANCED DISEASE SUSCEPTIBILITY TO ERYSIPHE ORONTII 16, F25A4.31, ICS1, ISOCHORISMATE SYNTHASE, ISOCHORISMATE SYNTHASE 1, SALICYLIC ACID INDUCTION DEFICIENT 2, SID2
Chromosome Location
Chr.1: 28070295 - 28074177 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106129.4 NP_565090.1 2 - 3 398 67
NM_202414.1 NP_974143.1 2 - 3 397 67
GenBank mRNA AF367342.1 - 2 - 3 396 67
AY056055.1 - 2 - 3 398 67
AY124864.1 - 2 - 3 302 67

View Full Product Details