Assay Design



Gene Transcripts

Gene Symbol
GH9B7
Entrez Gene ID
843902
Gene Name
glycosyl hydrolase 9B7
Gene Aliases
AT1G75680, AtGH9B7, F10A5.13, F10A5_13, glycosyl hydrolase 9B7
Chromosome Location
Chr.1: 28417052 - 28419635 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106219.3 NP_177697.1 2 - 3 905 95
GenBank mRNA AY039938.1 - 2 - 3 801 95
AY150451.1 - 2 - 3 501 95

View Full Product Details