Assay Design
Gene Transcripts
- Gene Symbol
- sks5
- Entrez Gene ID
-
843948
- Gene Name
- SKU5 similar 5
- Gene Aliases
-
AT1G76160, SKU5 similar 5, T23E18.10, T23E18_10
- Chromosome Location
-
Chr.1: 28577842 - 28581315 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_106265.4 |
NP_177743.1 |
4 - 5 |
918 |
93 |
GenBank mRNA |
AK316846.1 |
- |
4 - 5 |
711 |
93 |
View Full Product Details