Assay Design
Gene Transcripts
- Gene Symbol
- HMG1
- Entrez Gene ID
-
843982
- Gene Name
- hydroxy methylglutaryl CoA reductase 1
- Gene Aliases
-
3-HYDROXY-3-METHYLGLUTARYL COA REDUCTASE, 3-HYDROXY-3-METHYLGLUTARYL COA REDUCTASE 1, AT1G76490, AtHMGR1, F15M4.1, HMG-COA REDUCTASE, HMGR1, MAD3, hydroxy methylglutaryl CoA reductase 1, miRNA action deficient 3
- Chromosome Location
-
Chr.1: 28695760 - 28698852 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_106299.4 |
NP_177775.2 |
1 - 2 |
1176 |
127 |
GenBank mRNA |
AY488113.1 |
- |
1 - 2 |
1176 |
127 |
View Full Product Details