Assay Design
Gene Transcripts
- Gene Symbol
- CP12-3
- Entrez Gene ID
-
843989
- Gene Name
- CP12 domain-containing protein 3
- Gene Aliases
-
AT1G76560, CP12 domain-containing protein 3, F14G6.16, F14G6_16
- Chromosome Location
-
Chr.1: 28728206 - 28728976 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_106306.3 |
NP_565134.1 |
1 - 1 |
593 |
102 |
GenBank mRNA |
AK227842.1 |
- |
1 - 1 |
540 |
102 |
View Full Product Details