Assay Design
Gene Transcripts
- Gene Symbol
- PRAF1
- Entrez Gene ID
-
844030
- Gene Name
- Regulator of chromosome condensation (RCC1) family with FYVE zinc finger domain-containing protein
- Gene Aliases
-
AT1G76950, F22K20.5, F22K20_5
- Chromosome Location
-
Chr.1: 28906441 - 28911584 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_106346.3 |
NP_565144.1 |
4 - 5 |
829 |
116 |
GenBank mRNA |
AF323270.1 |
- |
4 - 5 |
783 |
116 |
View Full Product Details