Assay Design
Gene Transcripts
- Gene Symbol
- CYCA1;2
- Entrez Gene ID
-
844075
- Gene Name
- CYCLIN A1;2
- Gene Aliases
-
AT1G77390, CYCA1, CYCLIN A1, CYCLIN A1;2, DYP, F2P24.10, F2P24_10, TAM, TARDY ASYNCHRONOUS MEIOSIS
- Chromosome Location
-
Chr.1: 29081654 - 29084285 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_106388.4 |
NP_177863.2 |
1 - 2 |
312 |
106 |
View Full Product Details