Assay Design



Gene Transcripts

Gene Symbol
KOM
Entrez Gene ID
844122
Gene Name
Rhomboid-related intramembrane serine protease family protein
Gene Aliases
AT1G77860, F28K19.7, F28K19_7, KOMPEITO
Chromosome Location
Chr.1: 29282162 - 29284480 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334801.1 NP_001321009.1 3 - 4 662 99
NM_106435.2 NP_177909.2 3 - 4 640 99

View Full Product Details