Assay Design



Gene Transcripts

Gene Symbol
PUM7
Entrez Gene ID
844152
Gene Name
pumilio 7
Gene Aliases
APUM7, AT1G78160, T11I11.10, T11I11_10, pumilio 7
Chromosome Location
Chr.1: 29407608 - 29410484 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334820.1 NP_001322141.1 3 - 4 1581 78
NM_106466.2 NP_177940.2 2 - 3 1435 78

View Full Product Details