Assay Design
Gene Transcripts
- Gene Symbol
- VHA-A
- Entrez Gene ID
-
844228
- Gene Name
- vacuolar ATP synthase subunit A
- Gene Aliases
-
AT1G78900, vacuolar ATP synthase subunit A
- Chromosome Location
-
Chr.1: 29660088 - 29665078 on Build TAIR10.1
- Gene Symbol
- AT5G21020
- Entrez Gene ID
-
832226
- Gene Name
- transmembrane protein
- Gene Aliases
-
AT5G21020
- Chromosome Location
-
Chr.5: 7138762 - 7139661 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK230328.1 |
- |
- |
1538 |
67 |
View Full Product Details