Assay Design



Gene Transcripts

Gene Symbol
LUP2
Entrez Gene ID
844236
Gene Name
lupeol synthase 2
Gene Aliases
AT1G78960, ATLUP2, YUP8H12R.43, YUP8H12R_43, lupeol synthase 2
Chromosome Location
Chr.1: 29696394 - 29701439 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334861.1 NP_001322959.1 11 - 12 1899 91
NM_001334862.1 NP_001322960.1 12 - 13 1665 91
NM_001334863.1 NP_001322961.1 11 - 12 1823 91
NM_001334864.1 NP_001322958.1 10 - 11 1393 91
NM_106545.4 NP_178017.2 12 - 13 1694 91
GenBank mRNA AY091131.1 - 12 - 13 1618 91
AY114039.1 - 11 - 12 1571 91

View Full Product Details