Assay Design



Gene Transcripts

Gene Symbol
MC6
Entrez Gene ID
844270
Gene Name
metacaspase 6
Gene Aliases
AT1G79320, AtMC6, AtMCP2c, MCP2c, YUP8H12R.6, YUP8H12R_6, metacaspase 2c, metacaspase 6
Chromosome Location
Chr.1: 29836644 - 29838140 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106580.2 NP_178050.1 1 - 2 394 111
GenBank mRNA AY219831.1 - 1 - 2 352 111
AY322532.1 - 1 - 2 352 111
BT029232.1 - 1 - 2 352 111

View Full Product Details